VWR® Talon® Adjustable Cylinder Stand
Supplier: VWR International
97035-540
97035-540EA 806.83 USD
97035-540
VWR® Talon® Adjustable Cylinder Stand
Support Stands
Heavy-duty, cast aluminum stand with adjustable L-brackets holds cylinders from 15.2 – 23.5 cm (6 – 91/4") in diameter.
  • Prevents accidental tipping
  • Fully adjustable stand
  • Holds a range of cylinder sizes
Hinges open for installation without lifting the cylinder or disturbing connections.