VWR® Heavy-Duty Bench Clamp
Supplier: VWR International
97035-532
97035-532EA 136.54 USD
97035-532
VWR® Heavy-Duty Bench Clamp
Clamps General Purpose Clamps
Rugged, cast aluminum bench clamp has two screw clamps to tighten for extra firm mounting to any flat surface up to 4.5 cm (13/4") thick with a 3.2 cm (11/4”) overhang.
  • Aluminum construction
  • Convenient for temporary storage
  • Nickel-plated, non-slip spring catch buckle
Especially convenient for temporary storage situations. Features a 2.5 cm (1") wide, 137.2 cm (54") long nylon strap with nickel-plated, non-slip spring catch buckle to hold cylinders from 10.2–35.6 cm (4–14”) in diameter.
Ordering information: Not available with a safety message strap.