Excellence MP50 Benchtop Melting Point System, METTLER TOLEDO®
Supplier: Avantor
97035-162EA 11892.93 USD
97035-162
Excellence MP50 Benchtop Melting Point System, METTLER TOLEDO®
Melting Point Apparatus
The METTLER TOLEDO® MP50 benchtop melting point system gives you the ability to record and review melting episodes for investigation of unexpected results.
  • Record and review melting episodes
  • Unlimited grayscale viewing
  • LED light source and built-in camera
  • Sophisticated and intuitive software
Unlimited grayscale viewing directly on the MP50’s integrated video screen with up to 6x magnification enable discrimination powerful enough to observe individual crystals of your preparations melt
The MP50 utilizes an LED light source and built-in camera to capture and automatically measure both reflected and transmitted light so the color of your sample will not compromise results. This innovative measuring principle combined with the ability to review melting episodes saves you time and money through elimination of obvious outliers caused by events such as decomposition.
The MP50’s sophisticated yet intuitive software enables you to easily perform complex statistical evaluation methods so you have full confidence in your data. One Click® shortcuts streamline workflows by allowing you to initiate your most frequent methods via a single icon tap on the color touchscreen. The MP50’s elegant ergonomic design, customizable user settings, and multilingual support (English, Spanish, Chinese, German, and Japanese) make the MP50 appealing and easy to use by a wide variety of operators.
The available MP Accessory Kit contains everything you will need to get started including five sample preparation tools, mortar, pestle, capillaries, forceps, and calibration standards with certificates in a reusable hard-shell case.
Recording also affords you the freedom to perform other important duties while your samples run.
Certifications: METTLER TOLEDO® provides ISO 9001 manufacturing, nationwide ISO 9001 field service, ISO 9001 quality certification, and ISO 14001 environmental certification. CE conformity mark provides you with the assurance that our products comply with the most recent EU directives. This product is CSA certified and UL Listed.
Accessories information: Accessories and reference standards are also available. Contact your Avantor sales representative for more information.