RS-P26 Printer, METTLER TOLEDO®
Supplier: Mettler Toledo
11124323
97034-192EA 1676.31 USD
97034-192
RS-P26 Printer, METTLER TOLEDO®
Printers Document Printers
The RS-P26 Compact Printer generates GxP-compliant printouts on regular or adhesive paper.
  • Compact design
  • Ergonomic
  • Easy operation
  • Allows taring and printing of balance
The ergonomic design makes operation easy and allows taring and printing of the balance directly from the printer.
The large paper rolls allow printing with fewer refills. The Printer automatically detects the settings of the connected device. Thanks to this feature no manual setting changes are required. The build-in real time clock and calendar combined with the lightfast paperMore Product Information