Glove Box, Hypoxia Chamber, 856-HYPO, Plas-Labs™
Supplier: Plas-Labs
856-HYPO
97034-150EA 43553.53 USD
97034-150
Glove Box, Hypoxia Chamber, 856-HYPO, Plas-Labs™
Glove Boxes and Glove Bags
856-HYPO, Hypoxia Chamber Glove Box
This new Hypoxia Chamber Glove Box is ideal for tissue culture work, including tumor cell and stem cell research. It features a color operated "touch screen" for automatically controlling oxygen levels, temperature levels, and carbon dioxide levels.
Certifications: UL Listed.