Ion exchange Chromatography Columns, BioPro Series, YMC America
Supplier: YMC America,Inc
SF00S05-1046WP
97032-596EA 2186.72 USD
97032-596
Ion exchange Chromatography Columns, BioPro Series, YMC America
Chromatography Columns
YMC-BioPro ion exchange columns are used for the separation of proteins, peptides, and nucleic acids.
  • The non-porous particles offer high efficiency and exceptional resolution
YMC-BioPro IEC columns are available in QA and SP chemistries and are based on porous (QA and SP columns) and non-porous (QA-F and SP-F columns) hydrophilic polymer beads with low nonspecific adsorption.