HPLC Columns, YMC-Pack PVA-Sil
Supplier: YMC America,Inc
PV12S05-2546WT
97032-446EA 1670.47 USD
97032-446
HPLC Columns, YMC-Pack PVA-Sil
Chromatography Columns
PVA-Sil, polyvinyl alcohol (PVA) surface chemistry, is an alternative to silica gel or other polar bonded phases which are used in normal phase chromatography.
  • Resolve compounds that behave poorly on silica
  • Alcohol functionality present on PVA-Sil is better suited for troublesome compounds
Characteristics and a unique selectivity often resolve compounds that behave poorly on silica. The alcohol functionality present on PVA-Sil is better suited for troublesome compounds, such organic bases, than acidic silanols present in unbonded silica.