YMC-Pack J'sphere M80 HPLC Columns, YMC America
Supplier: YMC America,Inc
JM08S04-1546WT
97031-252EA 1662.63 USD
97031-252
YMC-Pack J'sphere M80 HPLC Columns, YMC America
Chromatography Columns
J’sphere ODS packings are based on a spherical, ultra pure, high surface area silica which has a very homogeneous surface providing additional siloxane groups.
  • High quality RP columns
  • Polarity range created solely by C18 bonding density
  • Metabolite recognition
  • High siloxane content
  • Additional selectivity through H-bonding
The lower coverage of C18 monomeric bonded groups in J’sphere ODS-M80 provides moderate hydrophobicity. As the lipophilic character is decreased, the H-bonding capacity becomes important. J’sphere ODS-M80 has a pronounced balanced polarity which is flexible and allows application to a wide variety of separation problems. These columns are used in various fields in analytical HPLC including drug analysis and QC.