HPLC Columns, YMC Carotenoid
Supplier: YMC America,Inc
CT99S03-1002WT CT99S03-1003WT CT99S03-1046WT CT99S03-1502WT CT99S03-1503WT CT99S03-1546WT CT99S03-2502WT CT99S03-2546WT CT99S05-0520WT CT99S05-1002WT CT99S05-1046WT CT99S05-1502WT CT99S05-1503WT CT99S05-1520WT CT99S05-1546WT CT99S05-2502WT CT99S05-2503WT CT99S05-2510WT CT99S05-2520WT CT99S05-2546WT CT99S03-0204GC CT99S03-0502WT CT99S03-0503WT CT99S03-0504WT CT99S03-0546WT CT99S05-01Q1GC CT99S05-0204GC CT99S05-02Q1GC CT99S05-0502WT CT99S05-0503WT CT99S05-0504WT CT99S05-0546WT
97030-658EA 1354.41 USD
97030-658 97030-660 97030-662 97030-664 97030-666 97030-668 97030-670 97030-672 97030-694 97030-698 97030-700 97030-702 97030-704 97030-706 97030-708 97030-710 97030-712 97030-714 97030-716 97030-718 89216-656 89216-660 89216-662 89216-664 89216-666 89216-668 89216-670 89216-672 89216-674 89216-676 89216-678 89216-680 10811-238
HPLC Columns, YMC Carotenoid
Chromatography Columns
The Carotenoid C30 stationary phase provides sufficient phase thickness to enhance interaction with long chain molecules therefore, geometric and positional isomers of conjugated double-bonding systems are recognized and resolved.
  • These columns is much more hydrophobic and lipophilic
  • Even when pure organic eluents are applied, many sample solutes are retained
  • Applicable for LC-MS
Carotenoids are found in a variety of natural sources including fruits and vegetables. In addition, carotenoids are considered as potential drugs for cancer intervention and/or prevention. Despite the complexity of carotenoid extracts and the minor shape differences between carotenoid isomers, the separation, identification, and quantitation of these compounds can be achieved using these columns.