Cimarec™ Basic Hotplates, Thermo Scientific
Supplier: Thermo Fisher Scientific
HP194515 HP194825
97027-350EA 288.8 USD
97027-350 97027-356
Cimarec™ Basic Hotplates, Thermo Scientific
Heaters Hot Plates
A sleek, rugged design and construction reduces the chance of top plate breakage and promotes durability, for economical high performance.
  • Reach maximum temperature with in 8 minutes
  • Solid ceramic top plate cleans easily, resists acids and alkalis
  • New top-plate design reduces chance of breakage
  • Spillaway design diverts spills away from internal components
  • Control knob allows precise temperature control
  • Integral ring-stand holder accommodates 1.3 cm (1/2") diameter support rod