Hardness 1 Buffer Solution, Hach
Supplier: Hach
42453
97026-632EA
130.81
USD
97026-632
97026-664
97026-364
Hardness 1 Buffer Solution, Hach
Environmental Test Kits
Buffer for hardness and chelant determination by titration, pH 10.1 ± 0.1 at 20°C
Hach methods 8213 and 8226. Also used in Hach Phenols method 8047.