COD Digestion Vials, Manganese III, Hach
97026-430PK
122.89
USD
97026-430
COD Digestion Vials, Manganese III, Hach
Environmental Test Kits
For determination of Chemical Oxygen Demand (COD) by the Manganese III Reactor Digestion Method
Hach Method 10067.
Ordering information: Pack of 25 (pack of 150 vials available). Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)