InLab® 605 Dissolved Oxygen Sensors, METTLER TOLEDO®
Supplier: Mettler Toledo
51340291 51340292 51344611 51344613
11278-380EA 1390 USD
11278-380 11278-382 97025-894 97025-896
InLab® 605 Dissolved Oxygen Sensors, METTLER TOLEDO®
Electrodes Dissolved Oxygen Electrodes
Sensors ensure a high degree of accuracy and reproducibility for polarographic dissolved oxygen determination.
  • High degree of accuracy
  • For polargraphic DO determination
  • Withstands harsh conditions
  • Steel mesh protects membrane
  • Reinforced shaft
Designed to withstand the harshest of conditions, sensors feature a membrane protected by steel mesh, a shaft constructed from glass fiber reinforced polyphenylene sulfide (PPS), and BNC/Cinch connectors. For use with SevenGo® portable dissolved oxygen meters. Sensors meet full IP67 protection standards.
Ordering information: Sensors with ISM® technology also available. Accessories sold separately.