SevenGo Duo Pro™ SG78 Portable pH/Ion/Conductivity Meter, METTLER TOLEDO®
Supplier: Mettler Toledo
51302622 51302625 51302621 51302620
97025-866EA 3149.18 USD
97025-866 97025-868 97025-864 97025-862
SevenGo Duo Pro™ SG78 Portable pH/Ion/Conductivity Meter, METTLER TOLEDO®
Electrochemistry Meters pH and Multi-Parameter Meters
Ideal for routine pH, mV, Conductivity, Salinity, Ion concentration, Specific resistance and TDS measurements
  • Multiparameter
  • Store up to 500 data points
  • Automatic endpoint and buffer recognition
  • Temperature compensation
  • Five-point calibration
These units store up to 500 GLP data points with Time/Date, Sensor ID, User name, and Sample ID. Units feature automatic endpoint and buffer recognition, both linear and non-linear temperature compensation, and five-point pH calibration with seven predefined and one user-defined buffers. It is equipped with sophisticated new concept, Intelligent Sensor Management (ISM®). The display shows measured values and settings while continuously indicating electrode and battery status. Units are equipped with rotatable electrode clip that can be mounted on the left or the right. It allows single-handed measurement and saves space when the electrode is not being used.
Certifications: Meter meets IP67 specifications, allowing the sensor to be immersed in water to a depth of 1m for 1 hour without any water incursion.
Ordering information: All meters come with a wrist strap, four AA batteries, instruction manual, quality certificate, and declaration of conformity. Electrode kit and Field kit versions include all items as the instrument only version plus InLab® Expert-Pro-ISM and InLab® 738-ISM (both IP 67 and 1.8m fixed cable). USP/EP kit includes all items as Electrode kit, plus different sensors such as Inlab® Pure Pro-ISM, 2m ISM-electrode cable and Inlab® 742-ISM. In addition Field kit includes uGo™ field compact case, rubber holster, 2 SevenGo clips, buffer sachets pH (2 each: 4.00, 7.00), conductivity standard sachets (2 each; 1413 µS/cm, 12.88 mS/cm) and calibration bottles.