Benchtop Densitometer Turbidimeter, McFarland Units, DEN-1, Grant Instruments
Supplier: GRANT USA INC.
DEN-1
97025-504EA 1273 USD
97025-504
Benchtop Densitometer Turbidimeter, McFarland Units, DEN-1, Grant Instruments
Turbidity Meters
The Grant bio DEN-1 is a compact densitometer for measuring turbidity of cell suspensions in a range of life science applications, ideal for measurements on the bench or in cold rooms, incubators and workstations.
Small, lightweight, easy to use instruments. Rapid, clear results displayed digitally in McFarland units. Can be user calibrated with commercial standards or cell suspensions prepared in the laboratory. Light diode light source.
Certifications: CE marked.
Ordering information: Instruments supplied with a D16 adapter for Ø 16 mm tubes. Manufacturers 2 year warranty.