Micro Plastic Cells, PerkinElmer
Supplier: PerkinElmer
L7110262
97024-690PK 315.55 USD
97024-690
Micro Plastic Cells, PerkinElmer
Cuvettes Spectrophotometer Cuvettes
These disposable micro cells have a volume of 70 to 550 ml, and a path length of 10 mm.
  • Cells are individually wrapped
Disposable cells greatly reduce the risk of contamination, removing the need for cleaning reagents and reducing the laboratory costs when compared to quartz or glass cells. Each cell is suitable for any UV/Vis spectrophotometer which accommodates cells with a center height of 15 mm.