Micro Plastic Cells - 10 mm, PerkinElmer
Supplier: PerkinElmer
L7110260
97024-686PK 991.15 USD
97024-686
Micro Plastic Cells - 10 mm, PerkinElmer
Cuvettes Spectrophotometer Cuvettes
These disposable micro cells have a volume of 70-550 µl, a pathlength of 10 mm and outside dimensions of 45 mm (H)×12.5 mm (W)×12.5 mm (D).
Disposable cells greatly reduce the risk of contamination, removing the need for cleaning reagents and reducing the laboratory costs as compared to quartz or glass cells. Each cell is suitable for any UV/Vis spectrophotometer which accommodates cells with a center height of 15 mm.