Beam Condenser for FT-IR, PerkinElmer
Supplier: PerkinElmer
01860794
97024-636EA 7358.18 USD
97024-636
Beam Condenser for FT-IR, PerkinElmer
Spectrophotometers and Photometers IR and FTIR Spectrophotometers
A beam condenser is required for microsamples such as KBr microdisks, paint flakes, small spots on polymer films, and even single fibers when an optional fiber holder is used.
  • Optimum light-gathering power
  • Easy measurement of microgram and submicrogram quantities of sample
  • Highly efficient optical design results in high-quality FT-IR spectra
Delivery information: Includes the transfer optics and Microsample Disk Holder. Optional Miniature Diamond Anvil Cell™ is available for high-pressure analysis of small, optically thick polymeric samples.