Variable Angle Reflectance Accessory for PerkinElmer Lambda Spectrometers, PerkinElmer
Supplier: PerkinElmer
B0137314
97024-530EA 10476.92 USD
97024-530
Variable Angle Reflectance Accessory for PerkinElmer Lambda Spectrometers, PerkinElmer
Spectrophotometers and Photometers UV/Visible Spectrophotometers
The variable angle reflectance accessory is for use with LAMBDA 25/35/45/650/750/850/950/1050. The variable angle reflectance accessory is ideal for relative specular reflectance studies which require data from more than one angle of incidence.
  • Wavelength range of 175 to 3300 nm and locates in standard sample compartment
  • Angular range from 15 to 70° with an accuracy of ±1°
Typical applications include the study of how the reflectivity of a sample changes with change of incident angle.