OhausⓇ Moisture Analyzers, MB Series
Supplier: Ohaus
83999801
97021-958EA 4314.57 USD
97021-958
OhausⓇ Moisture Analyzers, MB Series
Analyzers Moisture Analyzers
Model MB23 basic moisture analyzers combine high quality durable construction with easy one-button setup, ideal for operators of all skill levels
Feature fast and accurate results, large backlit LCD display, standard RS232 port, readabilities from 0.1% to 0.05%, compact footprint, and a choice of halogen or infrared heating
Certifications: UL listed. CSA certified. FCC approved. CE marked. Manufactured under an ISO 9001:2000 Registered Quality Management System.
Ordering information: Manufacturer’s two-year parts and labor warranty (U.S. and Canada).