ASTM Class 1 Weight Sets, METTLER TOLEDO®
Supplier: Mettler Toledo
11124002 11124008 11124018 11124005 11124015 11124019 11124000 11124004 11124014 11124001 11124003 11124006 11124009 11124007
97021-840EA 1795 USD
97021-840 97021-846 97021-850 97021-856 97021-862 97021-864 97021-838 97021-842 97021-848 97021-852 97021-854 97021-844 97021-860 97021-858
ASTM Class 1 Weight Sets, METTLER TOLEDO®
Weights Calibration Weights
These ASTM weight sets with adjusting cavity are made of vaccum-melted stainless steel, with an electropolished finish.
  • Electrolytic polishing ensures glossy surfaces for anti-adhesion effects
  • Exclusive material has a density of 7.9 kg/dm3