Column Cutters, PerkinElmer
Supplier: PERKINELMER U.S. LLC
N9301376
97019-380PK 192.15 USD
97019-380
Column Cutters, PerkinElmer
Cutters
This ceramic column cutter is ideal for cutting polyimide fused silica capillary columns and guard columns.
  • Easy to hold and simple to use
  • All four sides can be used as a cutting tool