Xenon Lamps for Fluorescence Detectors, PerkinElmer
Supplier: PerkinElmer
N2922082
97019-248EA 4064.7 USD
97019-248
Xenon Lamps for Fluorescence Detectors, PerkinElmer
Lights Xenon Lights, Lamps and Bulbs
The Xenon source (150 W) used in Series 200 Fluorescence Detector provides signal to noise ratio of greater than 700:1 from trace analysis.