Cold Traps, PerkinElmer
Supplier: PerkinElmer
N6200150 L4275110 L1003028 N6301054 L4275009 L4271187 M0413535 M0413627 L4271290 L4271203 L4071151 L4271106 L4271205 N9306093 N9306094 N9306096 L1003027 M0413628 L4275107 L4270123 L4275108 N6101474 N9306095
97019-322EA 1262.41 USD
97019-322 97012-816 97012-818 97013-012 97013-034 97013-036 97013-154 97013-156 97013-638 97013-846 97019-146 97019-158 97019-160 97019-624 97019-626 97019-630 97012-814 97013-140 97013-144 97013-712 97013-840 97019-164 97019-306 97019-628
Cold Traps, PerkinElmer
Vacuum Traps
TurboMatrix thermal desorption trap supplies exceptional analytical performance, used for U.S.
EPA Method TO17, the PerkinElmer standard trap, packed with Tenax™, on the TurboMatrix™ thermal desorber, will improve productivity and trapping capacity. The TurboMatrix air monitoring trap is packed with carbonaceous sorbents suitable for ozone precursor and air toxics monitoring.