Borosilicate Glass Sample Tubes, PerkinElmer
Supplier: PERKINELMER U.S. LLC
L4071594
97019-148PK 751.4 USD
97019-148
Borosilicate Glass Sample Tubes, PerkinElmer
Chromatography Supplies Chromatography Thermal Desorber Tubes
Empty borosilicate glass sample tubes without caps for industrial hygiene applications using PerkinElmer thermal desorption (TD) instruments.
The tubes are a component in the workplace air monitoring industrial hygiene Application Kit (N6100448) for thermal desorption instruments.