RS-P25 Printer, METTLER TOLEDO®
Supplier: Mettler Toledo
30702967
97016-768EA 1192.15 USD
97016-768
RS-P25 Printer, METTLER TOLEDO®
Printers Document Printers
The RS-P25 Compact Printer provides simple printouts on lightfast or adhesive paper.
  • Fast and reliable printer
  • User friendly operation
  • RS232 interface
For use with certain METTLER TOLEDO® balances, titrators, and Ph meters.