Design-Aire Fan Filter Modules, Design Filtration
Supplier: Microzone
DA2-1HSL DA4-1HSL DA3-1HSL DA3-2HSL DA4-2HSL
97015-388EA 3972.25 USD
97015-388 97015-392 97015-390 97015-402 97015-404
Design-Aire Fan Filter Modules, Design Filtration
Prefabricated Rooms
Standard features include room-side replaceable filter, durable aluminum housing, disposable prefilters rated MERV 7, variable speed control (supplied loose with module not installed), duplex electrical outlet (15 A, 115 V or 230 V), and 15' power cord (115 V or 230 V).
HEPA Filters are rated 99.99% efficient at 0.3 Microns; ULPA Filters are rated 99.9995% efficient at 0.12 Microns.