N920 Diapragm Vacuum Pump, KNF
Supplier: KNF Neuberger Inc.
302473/057334
97014-794EA 9156.28 USD
97014-794
N920 Diapragm Vacuum Pump, KNF
Pumps Vacuum Diaphragm Pumps
Diaphragm stabilization system allows high suction speed even in the low-vacuum range.
  • Features an integrated potentiometer to ensure desired pumping rate selecting appropriate speed
  • 100% pure oil-free transfer, evacuation and compression without contaminating the medium
  • With PPS pump head, PTFE coated diaphragm, FFPM valves
  • Three stage pump head improve leak-tightness
  • Gas tight, leakage rate: 6×103 mbar
  • Maintenance-free, environmentally friendly pump with adjustable flow rate and thermal switch
  • Universal power requirement
  • For slightly aggressive or corrosive gases and vapors
Optional analog signal input available for external control.