Pulp and Paper Blotting Paper, Grade 240, Ahlstrom
Supplier: Ahlstrom
2400-0808
97014-664CS 600.04 USD
97014-664
Pulp and Paper Blotting Paper, Grade 240, Ahlstrom
Blotting Paper
Ahlstrom Grade 240 is recommended as a blotter that meets standards for use in TAPPI T-205 and T-491.