Pressure Regulator Fixtures, WaterSaver Faucet
Supplier: WaterSaver Faucet
L2873-771WSA
97014-324EA 1112.54 USD
97014-324
Pressure Regulator Fixtures, WaterSaver Faucet
Gas Regulators
Pressure regulator used for compressed air and inert high purity gases, including nitrogen, helium, argon and oxygen.
  • 3/8" NPT male inlet
  • Removable quick connect fitting with 1/4" NPT male plug
  • Non-relieving type with brass body and neoprene diaphragm
Fine control needle valve for precise flow control. Valve has forged brass body, floating tapered stainless steel needle and replaceable stainless steel seat.