Cyclonic/Concentric Sample Introduction Torch Module for Optima™ 2x00/4x00/5x00/7x00 DV, PerkinElmer
Supplier: PerkinElmer
N0770607
97013-832EA 6439.62 USD
97013-832
Cyclonic/Concentric Sample Introduction Torch Module for Optima™ 2x00/4x00/5x00/7x00 DV, PerkinElmer
Chromatography Supplies Chromatography Spare Parts
This quick-change sample introduction module, with cyclonic spray chamber and MEINHARD® concentric nebulizer technologies, includes core consumables and components for Optima™ 2x00 DV, 4x00 DV, 5x00 DV, and 7x00 DV ICP-OES series instruments.
Delivery information: This kit includes: (1) MEINHARD® Type C Concentric Glass Nebulizer, (1) 1-Slot Quartz Torch, (1) Cyclonic Spray Chamber Assembly, (1) Quick-Change Torch Module Mounting Block, (1) 2.0 mm Int.Ø Alumina Injector, (1) Nebulizer Tubing Assembly, and (1) Polyethylene Tubing (Int.Ø×L: 0.58 mm × 25.4 cm)