Septum Cap for Auto SYS™ XL, PerkinElmer
Supplier: PERKINELMER U.S. LLC
N6100153
97013-790EA 187.18 USD
97013-790
Septum Cap for Auto SYS™ XL, PerkinElmer
Septa
Septum cap made of clear anodized aluminum and high-quality stainless steel.
  • Highly durable
  • Meet or exceed manufacturer's performance standards