Ryton® Scott Spray Chamber for Optima 3000 DV/3000 SCR/3×00 RL, PerkinElmer
Supplier: PerkinElmer
N0690640
97013-782EA 773.98 USD
97013-782
Ryton® Scott Spray Chamber for Optima 3000 DV/3000 SCR/3×00 RL, PerkinElmer
Chromatography Supplies Chromatography Spare Parts
The Ryton® scott spray chamber is designed for use with Optima™ 3000, 3000 SCR, and 3×00 RL ICP-OES series instruments.
  • Chamber is made from corrosion-resistant Ryton®
  • Inert to most mineral acids including hydrofluoric acid, aqua regia, and all organic solvents normally used in ICP analyses
  • Scott-type design minimizes pulsations from the peristaltic pump
The type II, two-piece design ensures leak-free performance. The removable inner tube allows the chamber to be easily disassembled for cleaning.