Rectangular Semi-Demountable Cell Windows, PerkinElmer
Supplier: PerkinElmer
L1271192 L1271191 L1271193
97013-768PK 528.64 USD
97013-768 97013-770 97013-766
Rectangular Semi-Demountable Cell Windows, PerkinElmer
Specimen Mounting Supplies
Rectangular windows make the demountable cell becomes a semi-demountable cell. It can be used either as a demountable cell or as a sealed cell for high-viscosity liquids or mull samples.
  • Available in a variety of window materials
  • Windows come in pairs
Combining two different requirements into one cell allows a myriad of different sampling applications.
Ordering information: Semi-demountable Cell can be ordered with the universal demountable cell, windows and spacers.