Nebulizer Cleaning Wires, PerkinElmer
Supplier: PERKINELMER U.S. LLC
03030135
97013-702PK 73.91 USD
97013-702
Nebulizer Cleaning Wires, PerkinElmer
Tool Sets
Made of soft alloy copper, these cleaning wires are specially deburred to prevent scratching of the orifices and internal components of the nebulizers while cleaning.
  • Soft copper construction
  • Deburred
  • Provided in convenient packs of multiples
Use only PerkinElmer cleaning wires to clean nebulizers.