THGA Furnace Tube Inspection Mirror, PerkinElmer
Supplier: PERKINELMER U.S. LLC
B0851900
97013-696EA 260.51 USD
97013-696
THGA Furnace Tube Inspection Mirror, PerkinElmer
Mirrors
This mirror is designed to observe the sample during an THGA furnace temperature programmed run. By observing the sample during the run, the drying temperatures and times can be optimized.
Ordering information: HGA instrument mirrors are available separately.