High Pressure Sample Pans, PerkinElmer
Supplier: PerkinElmer
03190218 03190029 B0182901 B0182902
97012-540PK 570.91 USD
97012-540 97013-690 97013-784 97012-954
High Pressure Sample Pans, PerkinElmer
Trays/Dishes/Pots Laboratory Trays/Dishes
Pans contain the sample in a sealed environment throughout an experiment.
  • Pressure ranges from 24 to 150 bar
The sealing suppresses the vaporization of a solvent or contains a volatile reaction product, thereby eliminating the interfering effects of the heat of vaporization.
The stainless steel pans with covers and seals are reusable and includes 5 pans and 20 gold-plated seals.