Sapphire Injector for PerkinElmer Optima ICP-OES Instruments, PerkinElmer
Supplier: PerkinElmer
N0775247
97013-662EA 3955.03 USD
97013-662
Sapphire Injector for PerkinElmer Optima ICP-OES Instruments, PerkinElmer
Microinjectors Microinjectors, Programmable
This sapphire injector is for use with Optima® 2.0 mm I.D. Sapphire Injector for Optima® 2x00/4x00/5x00/7x00 DV ICP-OES series instruments.
Sapphire injectors are ideal for silicon applications.