High Sensitivity GemTip Nebulizer, PerkinElmer
Supplier: PerkinElmer
N0370393
97013-650EA 4002.81 USD
97013-650
High Sensitivity GemTip Nebulizer, PerkinElmer
Atomizers
This high sensitivity plastic body nebulizer is for use with AAnalyst® 100/200/300/400/700/800 and earlier AA series instruments. It is corrosion resistant and can be used to aspirate almost all solutions, even those solutions containing a high percentage of acid.
A complete GemTip High-Sensitivity plastic body nebulizer featuring Kalrez O-rings and plastic needle with sapphire tip. This nebulizer was designed to obtain optimum sensitivity, detection limits and precision with flame atomic absorption for both air-acetylene and nitrous oxide-acetylene flames.
This assembly includes: capillary tubing, Qty.1 (97012-048), fine adjustment nut, Qty.1 (101680-080), gland nut, Qty.1 (97012-744), PTFE tubing, Qty.1 (97012-040), capillary assembly for high-sensitivity GemTip nebulizers, Qty.1 (10138-028), O-Ring for capillary assembly, Qty.1 (101692-784), impact bead for high-sensitivity GemTip nebulizer, Qty.1 (97012-848), O-Ring for nebulizer body, Qty.1 (97012-136), body assembly for high-sensitivity nebulizer, Qty.1 (10138-024), locking nut, Qty.1 (101675-032), spring, Qty.1, connector nut, Qty.1 (101675-030), magnet holder assembly, Qty.1 (101681-786). This nebulizer is compatible with the PerkinElmer AAnalyst® 200/400*, AAnalyst®100/300/700/800, and Models 5100PC/5000/3300/3110/3100 AA instruments. Note: AAnalyst® 200/400 requires an end cap assembly (10138-018).