GemTip Cross-Flow II Nebulizer for Optima ICP-OES Instruments, PerkinElmer
Supplier: PerkinElmer
N0770546
97013-634EA 2090.56 USD
97013-634
GemTip Cross-Flow II Nebulizer for Optima ICP-OES Instruments, PerkinElmer
Atomizers
This GemTip™ Nebulizer is for use with Optima® 2x00/4x00/5x00/7x00 ICP-OES series instruments. It is an excellent general purpose nebulizer for the analysis of strong mineral acids (including HF) and samples with less than 5% dissolved solids.
It can routinely handle 50% (v/v) solutions of HCl, HNO₃, H₂SO₄, H₂PO₄, 20% (v/v) HF and 30% (v/v) NaOH. Up to 20% NaCl can be aspirated by the nebulizer for 1 hour without clogging.
This nebulizer uses a chemically resistant GemTip made of sapphire and ruby in a chemically resistant end cap (97013-636). It is included in the Quick-Change Torch Module with Scott Spray Chamber and GemTip Cross-Flow II Nebulizer for Optima 2x00/4x00/5x00/7x00 DV (10136-310) and the Scott/Cross-Flow II Sample Introduction Kit for Optima 2x00/4x00/5x00/7x00 DV (101692-222).