Concentric Nebulizer/Scott Spray Chamber End Cap Assembly for Avio and Optima, PerkinElmer
Supplier: PerkinElmer
N0680504
97013-600EA 1147.94 USD
97013-600
Concentric Nebulizer/Scott Spray Chamber End Cap Assembly for Avio and Optima, PerkinElmer
Atomizers
This end cap assembly, which connects concentric GemCone™ and similar nebulizers to Ryton Scott spray chambers, is used with Avio® 200/500 ICP-OES, and Optima® 2x00 DV/4x00 DV/5x00 DV/7x00 DV/8x00 ICP-OES series instruments.
Delivery information: This assembly includes: Nebulizer O-Ring (97012-786) (1), Knurled Screw (2), Insertion Ring (1), O-Ring, 34.29 mm I.Ø, (1), Nut (1), End Cap (1), End Cap Washer (1).