GemCone Low-Flow Nebulizer for Optima® 3X00, PerkinElmer
Supplier: PERKINELMER U.S. LLC
N0690671
97013-594EA 1277.93 USD
97013-594
GemCone Low-Flow Nebulizer for Optima® 3X00, PerkinElmer
Atomizers
This GemCone nebulizer is designed for use with the Optima® 3000 XL or the Optima® 3000 DV in the axial configuration. Low-flow GemCone Nebulizers permit lower nebulizer gas flow rates, useful for spectral lines with high excitation energies and for providing a more robust plasma.
GemCone Nebulizers require an end cap (N0680504) for use with Scott Spray Chambers.