Quartz Baffled Cyclonic Spray Chamber for ELAN DRC/DRCplus/DRC II, PerkinElmer
Supplier: PerkinElmer
WE025221
97013-558EA 2866.95 USD
97013-558
Quartz Baffled Cyclonic Spray Chamber for ELAN DRC/DRCplus/DRC II, PerkinElmer
Atomizers
This quartz baffled cyclonic spray chamber, with ball joint, is used for ELAN® DRC, DRCplus, and DRC II ICP-MS series instruments. It contains PTFE O-rings.
The spray chamber is an integral part of the sample introduction system responsible for altering the sample mist to permit the appropriate droplet size distribution to reach the plasma. The baffled cyclonic spray chamber features a central transfer tube which acts as a secondary particle separator helping separate larger aerosol particles from the sample. This reduces solvent load in the plasma without compromising detection limits.
The distance between the bottom of the baffle and the chamber wall is carefully controlled so that droplets cannot form on the bottom of the baffle. The advantage is smooth draining and improved precision.