Hand Crimper, PerkinElmer
Supplier: PerkinElmer
N9302785
97013-494EA 902.43 USD
97013-494
Hand Crimper, PerkinElmer
Crimpers, Cappers and Decappers
Hand crimper for GC Headspace vials or LC large volume vials.
  • Suited for large-volume vials
  • Guarantees sample integrity
  • Useful when handling volatile compounds
It ensures sample integrity, particularly those containing volatile compounds.