Single Stage High Purity Brass Regulators, PerkinElmer
Supplier: PERKINELMER U.S. LLC
00230090
97013-482EA 1124.74 USD
97013-482
Single Stage High Purity Brass Regulators, PerkinElmer
Gas Regulators
High purity brass regulators are constructed of high purity brass barstock and have stainless steel diaphragms and metal-to-metal seals. They are suitable for use with high purity (>99.995% pure) non-corrosive gases.
  • Regulators terminate in ¹/₄"
  • NPT swagelock fitting
Ideally suited for chromatographic carrier gas applications including FID, TCD,ECD, HID, and non-corrosive gas mixtures for analytical instrumentation.
Delivery information: Supplied with adapter.