Radial Purge Extension Window for PerkinElmer Optima ICP-OES Instruments, PerkinElmer
Supplier: PERKINELMER U.S. LLC
N0770944
97013-472EA 382.98 USD
97013-472
Radial Purge Extension Window for PerkinElmer Optima ICP-OES Instruments, PerkinElmer
Optical Windows
This radial view purge extension window is used with PerkinElmer Optima™ 5x00, 7100, 7200, 7300 DV, and 8300 ICP-OES series instruments.
The radial view purge extension window is a replaceable quartz window that slides into the mount between the torch and the optics for the radial view.