Radial Purge Extension Window for Optima ICP-OES Instruments, PerkinElmer
Supplier: PerkinElmer
N0581497
97013-462EA 732.27 USD
97013-462
Radial Purge Extension Window for Optima ICP-OES Instruments, PerkinElmer
Chromatography Supplies Chromatography Spare Parts
This quartz radial window for the purge extension is used with Optima™ 3000, 3000 SCR, 3x00 RL, 4300 V, 5300 V, and 7300 V ICP-OES series instruments.
The radial view purge extension window tube is a replaceable quartz tube that slides into the mount between the torch and the optics for the radial view.