Quartz SUPRASIL Macro/Semi-Micro Cell, PerkinElmer
Supplier: PerkinElmer
B0631132
97013-452EA 1498.46 USD
97013-452
Quartz SUPRASIL Macro/Semi-Micro Cell, PerkinElmer
Cuvettes Spectrophotometer Cuvettes
The Quartz SUPRASIL Macro/Semi-Micro Cell has a 4 x 4 mm light path with 4 windows.
  • 4 windows for excellent visibility
  • Interior width: 4 mm
  • Cell volume: 500 μL
Standard rectangular macro cells are the most frequently used type of spectroscopy cell for routine liquids analysis.
Ordering information: Cell requires stirrer fleas.