Disposable Reference Mirror for Reflectance Spectroscopy, Perkinelmer
Supplier: PerkinElmer
N1010504
97013-442EA 3347.4 USD
97013-442
Disposable Reference Mirror for Reflectance Spectroscopy, Perkinelmer
Mirrors
Reference mirror, disposable for reflectance spectroscopy, secondary standard for specular reflectance calibration according to GLP regulations, not recalibrated.