Universal Coded Intensitron Lamp Plug Adapter, PerkinElmer
Supplier: PerkinElmer
N3050196
97013-410EA 202.74 USD
97013-410
Universal Coded Intensitron Lamp Plug Adapter, PerkinElmer
Chromatography Supplies Chromatography Spare Parts
Allows coded lamps (N06612XX series, 12-pin) to be used with AAnalyst instruments (4-pin); however, an AAnalyst spectrometer does not recognize their coding.
For AAnalyst series of AA instruments with 4-pin lamp adapters.